Learn More
Eurofins Discoverx PathHunter NPY2R Bioassay Kit, 2-Plate. (93-0212Y2-00065)
Catalog No. 501999969
Encompass
The PathHunter NPY2R Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. peptide YY) based activation of NPY2R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged NPY2R and EA-tagged β-Arrestin-2. Activation of NPY2R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-199-9969 Supplier Eurofins Discoverx Supplier No. 930212Y200065
The PathHunter NPY2R Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. peptide YY) based activation of NPY2R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged NPY2R and EA-tagged β-Arrestin-2. Activation of NPY2R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.