Learn More
Eurofins Discoverx ChemiScreen Y2 Neuoropeptide Y Receptor Stable Cell Line, 2 vials. (HTS066C)
Catalog No. 502005790
Encompass
The ChemiScreen Y2 Neuropeptide Y Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. peptide YY) based activation of NPY2R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses NPY2R alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of NPY2R, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5790 Supplier Eurofins Discoverx Supplier No. HTS066C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen Y2 Neuropeptide Y Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. peptide YY) based activation of NPY2R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses NPY2R alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of NPY2R, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.