Learn More
Eurofins Discoverx Ready-to-Assay Y2 Neuropeptide Receptor Frozen Cells, 2 Vials of Cells. (HTS066RTA)
Catalog No. 502005792
Encompass
The Ready-to-Assay Y2 Neuropeptide Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. peptide YY) based activation of NPY2R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses NPY2R alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of NPY2R, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5792 Supplier Eurofins Discoverx Supplier No. HTS066RTA
The Ready-to-Assay Y2 Neuropeptide Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. peptide YY) based activation of NPY2R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses NPY2R alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of NPY2R, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.