Learn More
Eurofins Discoverx ChemiScreen Y4 Neuropeptide Y Receptor Stable Cell Line, 2 vials. (HTS204C)
Catalog No. 502006048
Encompass
The ChemiScreen Y4 Neuropeptide Y Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. peptide YY) based activation of NPY4R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses NPY4R alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of NPY4R, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-6048 Supplier Eurofins Discoverx Supplier No. HTS204C
Add to Cart
Add to Cart
Request a Quote
The ChemiScreen Y4 Neuropeptide Y Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. peptide YY) based activation of NPY4R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses NPY4R alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of NPY4R, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.