Learn More
Eurofins Discoverx ChemiScreen Y4 Neuropeptide Y Receptor Membrane Preparation, 1 vial. (HTS204M)
Catalog No. 502006049
Encompass
The ChemiScreen Y4 Neuropeptide Y Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of NPY4R surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-6049 Supplier Eurofins Discoverx Supplier No. HTS204M
The ChemiScreen Y4 Neuropeptide Y Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of NPY4R surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.