Learn More
Eurofins Discoverx Ready-to-Assay Y4 Neuropeptide Y Receptor Frozen Cells, 2 Vials of Cells. (HTS204RTA)
Catalog No. 502006050
Encompass
The Ready-to-Assay Y4 Neuropeptide Y Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. peptide YY) based activation of NPY4R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses NPY4R alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of NPY4R, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-6050 Supplier Eurofins Discoverx Supplier No. HTS204RTA
The Ready-to-Assay Y4 Neuropeptide Y Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. peptide YY) based activation of NPY4R GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses NPY4R alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of NPY4R, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.