Learn More
Darwin Chambers Company NQ16P PORTABLE INCUBATOR
SDP
Catalog No. NC1761508
Encompass_Preferred
NQ16PLUS Interior dimensions: 12.7" L x 9" W x 8.3" H. Exterior dimensions: 16.7"L x 12.8"W x 11.2"H. Weight: 6.64 lbs. NQ Series have several uses, such as: Blood sample transport, Water treatment research, and Maintaining microbiological cultures for cell research. Exterior construction: Double-wall foam insulation of high-density polyethylene, Foamed-in-place, Closed-cell insulation. Interior: Molded poly with coved floor for ease of cleaning. TEMP RANGE: AMBIENT +5C to 37C,1.0C TOLERANCE - CONTROL READOUT: ACTUAL AND SET-POINT VALUES - SENSOR: TYPE T THERMOCOUPLE - CONTROLLER: MICROPROCESSOR BASED - CALIBRATION CAPABILITY: 1.0C OFFSET INCREMENTS - CORD LENGTH: 6FT
Catalog No. NC1761508 Supplier Darwin Chambers Company Supplier No. NQ16
NQ16PLUS Interior dimensions: 12.7" L x 9" W x 8.3" H. Exterior dimensions: 16.7"L x 12.8"W x 11.2"H. Weight: 6.64 lbs. NQ Series have several uses, such as: Blood sample transport, Water treatment research, and Maintaining microbiological cultures for cell research. Exterior construction: Double-wall foam insulation of high-density polyethylene, Foamed-in-place, Closed-cell insulation. Interior: Molded poly with coved floor for ease of cleaning. TEMP RANGE: AMBIENT +5C to 37C,1.0C TOLERANCE - CONTROL READOUT: ACTUAL AND SET-POINT VALUES - SENSOR: TYPE T THERMOCOUPLE - CONTROLLER: MICROPROCESSOR BASED - CALIBRATION CAPABILITY: 1.0C OFFSET INCREMENTS - CORD LENGTH: 6FT