Learn More
Medchemexpress LLC HY-101054 5mg Medchemexpress, NQ301 CAS:130089-98-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1010545MG
Encompass_Preferred
Medchemexpress, HY-101054 5mg NQ301 CAS:130089-98-4 NQ301 is an antithrombotic agent; inhibits collagen-challenged rabbit platelet aggregation with an IC 50 of 10 mg/mL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2023