Learn More
Darwin Chambers Company NQ09 PORTABLE INCUBATOR
SDP
Catalog No. NC0917321
Encompass_Preferred
NQ09 Interior dimensions: 10.6" L x 7.5" W x 5.2" H. Weight: 4.7 lbs. NQ Series have several uses, such as: Blood sample transport, Water treatment research, and Maintaining microbiological cultures for cell research. Exterior construction: Double-wall foam insulation of high-density polyethylene, Foamed-in-place, Closed-cell insulation. Interior: Molded poly with coved floor for ease of cleaning. TEMP RANGE: AMBIENT +5C to 37C,1.0C TOLERANCE - CONTROL READOUT: ACTUAL AND SET-POINT VALUES - SENSOR: TYPE T THERMOCOUPLE - CONTROLLER: MICROPROCESSOR BASED - CALIBRATION CAPABILITY: 1.0C OFFSET INCREMENTS - CORD LENGTH: 6FT
Catalog No. NC0917321 Supplier Darwin Chambers Company Supplier No. NQ09
NQ09 Interior dimensions: 10.6" L x 7.5" W x 5.2" H. Weight: 4.7 lbs. NQ Series have several uses, such as: Blood sample transport, Water treatment research, and Maintaining microbiological cultures for cell research. Exterior construction: Double-wall foam insulation of high-density polyethylene, Foamed-in-place, Closed-cell insulation. Interior: Molded poly with coved floor for ease of cleaning. TEMP RANGE: AMBIENT +5C to 37C,1.0C TOLERANCE - CONTROL READOUT: ACTUAL AND SET-POINT VALUES - SENSOR: TYPE T THERMOCOUPLE - CONTROLLER: MICROPROCESSOR BASED - CALIBRATION CAPABILITY: 1.0C OFFSET INCREMENTS - CORD LENGTH: 6FT