Learn More
TARGETMOL CHEMICALS INC NQDI-1 25MG
SDP
Catalog No. 502580385
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NQDI-1 (NQDI 1) is a selective inhibitor of apoptosis signal-regulating kinase 1 (ASK1 MAP3K5) (IC50/Ki 3 uM/500 nM). purity: 97%
Catalog No. 50-258-0385 Supplier TARGETMOL CHEMICALS INC Supplier No. T310125MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NQDI-1 (NQDI 1) is a selective inhibitor of apoptosis signal-regulating kinase 1 (ASK1 MAP3K5) (IC50/Ki 3 uM/500 nM). purity: 97%