Learn More
Medchemexpress LLC HY-128895 5mg Medchemexpress, NQO1 activator 1 CAS:1800405-30-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1288955MG
Encompass_Preferred
Medchemexpress, HY-128895 5mg NQO1 activator 1 CAS:1800405-30-4 NQO1 activator 1 is an orally available NAD+ modulator. It reacts with NAD(P)H:quinone oxidoreductase 1 (NQO1) as a substrate, resulting in increases in intracellular NAD+ levels via NADH oxidation. Elevated NAD+ levels triggers the activation of SIRT1 and AMPK, and subsequently activates PGC-1α. NQO1 activator 1 improves energy metabolism and mitochondrial dysfunction in MELAS fibroblasts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9782