Learn More
Medchemexpress LLC NQO1 substrate 25mg | 2304503-05-5 | 25 MG
SDP
Catalog No. 5000229160
Encompass_Preferred
NQO1 substrate is a small-molecule research reagent that serves as a substrate for NAD(P)H:quinone oxidoreductase 1 (NQO1). It is reported as an efficient NQO1 substrate and is investigated for activity against NQO1-overexpressing, drug-resistant non-small-cell lung cancer. Supplied as a solid or as a 10 mM solution in DMSO for enzymology and cellular studies.
  • Acts as an efficient NQO1 substrate in enzymatic assays.
  • Reported antitumor activity in NQO1-overexpressing, drug-resistant NSCLC models.
  • Available as solid (5-200 mg) or as 10 mM in DMSO solution.
  • High purity suitable for research (98.0% by COA).
Catalog No. 50-002-29160 Supplier Medchemexpress LLC Supplier No. HY11431525MG
NQO1 substrate is a small-molecule research reagent that serves as a substrate for NAD(P)H:quinone oxidoreductase 1 (NQO1). It is reported as an efficient NQO1 substrate and is investigated for activity against NQO1-overexpressing, drug-resistant non-small-cell lung cancer. Supplied as a solid or as a 10 mM solution in DMSO for enzymology and cellular studies.
  • Acts as an efficient NQO1 substrate in enzymatic assays.
  • Reported antitumor activity in NQO1-overexpressing, drug-resistant NSCLC models.
  • Available as solid (5-200 mg) or as 10 mM in DMSO solution.
  • High purity suitable for research (98.0% by COA).