Learn More
Medchemexpress LLC NQTrp | 185351-19-3 | 98.4% | 360.36 | 5 MG
SDP
Catalog No. 5000400903
Encompass_Preferred
NQTrp is an aromatic naphthoquinone-tryptophan hybrid molecule that functions as an inhibitor of tau protein aggregation and exhibits generic anti-amyloidogenic effects. It effectively inhibits the in vitro aggregation of hexapeptide (41GCWMLY46 within the N-terminus of γD-crystallin) and full-length γD-crystallin.
  • Inhibits tau protein aggregation
  • Exhibits generic anti-amyloidogenic effects
  • Prevents the formation of elongated amyloid fibrils
  • Reduces cytotoxicity induced by aggregates in ARPE-19 cells
  • For research use only
Catalog No. 50-004-00903 Supplier Medchemexpress LLC Supplier No. HY197385MG
NQTrp is an aromatic naphthoquinone-tryptophan hybrid molecule that functions as an inhibitor of tau protein aggregation and exhibits generic anti-amyloidogenic effects. It effectively inhibits the in vitro aggregation of hexapeptide (41GCWMLY46 within the N-terminus of γD-crystallin) and full-length γD-crystallin.
  • Inhibits tau protein aggregation
  • Exhibits generic anti-amyloidogenic effects
  • Prevents the formation of elongated amyloid fibrils
  • Reduces cytotoxicity induced by aggregates in ARPE-19 cells
  • For research use only