Learn More
Aviva Systems Biology Corp NR1D2 Antibody - C-terminal region (ARP38568_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38568_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NR1D2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NR1D2. Alternate names: RVR, BD73, EAR-1RApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56707