Learn More
Aviva Systems Biology Corp NR1D2 Antibody - middle region (ARP33671_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33671_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR1D2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NR1D2. Alternate names: BD73, EAR-1r, HZF2, Hs.37288, RVR, EAR-1RApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55112