Learn More
Aviva Systems Biology Corp NR1H2 Antibody - middle region (ARP33481_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33481_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR1H2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NR1H2. Alternate names: LXR-b, LXRB, NER, NER-I, RIP15, UNRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55040