Learn More
Aviva Systems Biology Corp NR1H3 Antibody - N-terminal region (ARP38669_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38669_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NR1H3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NR1H3. Alternate names: LXRA, LXR-a, RLD-1ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Pig, Rabbit
Catalog No. 50-014-56747