Learn More
Aviva Systems Biology Corp NR1H5 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP37630_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nr1h5. It was validated on Western Blot. Nr1h5,nuclear receptor subfamily 1, group H, member 5,Fxrb; MGC141184; Nr1h5,nuclear receptor subfamily 1, group H, member 5,nuclear receptor subfamily 1, group H, member 5The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Fxrb, MGC141184ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-672