Learn More
Aviva Systems Biology Corp NR1I2 Antibody - C-terminal region (ARP39532_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39532_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NR1I2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NR1I2. Alternate names: BXR, PAR, PRR, PXR, SAR, SXR, ONR1, PAR1, PAR2, PARqApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57074