Learn More
Aviva Systems Biology Corp NR1I2 Antibody - N-terminal region (ARP36016_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36016_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR1I2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NR1I2. Alternate names: BXR, ONR1, PAR, PAR1, PAR2, PARq, PRR, PXR, SAR, SXRApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-55970