Learn More
Aviva Systems Biology Corp NR1I3 Antibody - middle region (ARP37876_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37876_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NR1I3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse NR1I3. Alternate names: CAR, MB67, Care2, ESTM32, AA209988, AI551208, CAR-betaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Mouse, Rat
Catalog No. 50-014-56454