Learn More
Aviva Systems Biology Corp NR1I3 Antibody - C-terminal region (ARP33674_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33674_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NR1I3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NR1I3. Alternate names: CAR, CAR-BETA, CAR-SV1, CAR-SV10, CAR-SV11, CAR-SV12, CAR-SV13, CAR-SV14, CAR-SV15, CAR-SV17, CAR-SV18, CAR-SV19, CAR-SV20, CAR-SV21, CAR-SV4, CAR-SV6, CAR-SV7, CAR-SV8, CAR-SV9, CAR1, MB67, MGC97144ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55114