Learn More
Aviva Systems Biology Corp NR1I3 Antibody - middle region (ARP45624_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45624_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR1I3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NR1I3. Alternate names: CAR, CAR1, MB67, MGC150433, MGC97144, MGC97209ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59077