Learn More
Aviva Systems Biology Corp NR2C1 Antibody - C-terminal region (P100812_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100812_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NR2C1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NR2C1. Alternate names: TR2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62722