Learn More
Aviva Systems Biology Corp NR2C1 Antibody - C-terminal region (ARP38287_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38287_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR2C1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human NR2C1. Alternate names: TR2, TR2-11ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-56597