Learn More
Aviva Systems Biology Corp NR2C1 Antibody - N-terminal region (ARP38286_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38286_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR2C1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NR2C1. Alternate names: TR2, TR2-11ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56596