Learn More
Aviva Systems Biology Corp NR2E1 Antibody - middle region (ARP33422_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33422_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR2E1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human NR2E1. Alternate names: TLL, TLX, XTLLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55014