Learn More
Aviva Systems Biology Corp NR2E1 Antibody - middle region (ARP45612_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45612_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR2E1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NR2E1. Alternate names: TLL, TLX, XTLLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59069