Learn More
Aviva Systems Biology Corp NR2F1 Antibody - N-terminal region (ARP45629_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45629_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR2F1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NR2F1. Alternate names: COUP-TFI, EAR-3, EAR3, ERBAL3, NR2F2, SVP44, TCFCOUP1, TFCOUP1ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-59081