Learn More
Aviva Systems Biology Corp NR2F6 Antibody - middle region (ARP32995_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32995_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NR2F6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NR2F6. Alternate names: EAR2, EAR-2, ERBAL2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-52882