Learn More
TARGETMOL CHEMICALS INC NR2F6 MODULATOR-1 5MG
SDP
Catalog No. 502579482
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NR2F6 modulator-1 is a potent modulator of nuclear receptor subfamily 2 group F member 6 (NR2F6). NR2F6 modulator-1 can be used to study immune regulation and modulation of tumour stem cell activity. purity: 97%
Catalog No. 50-257-9482 Supplier TARGETMOL CHEMICALS INC Supplier No. T622045MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NR2F6 modulator-1 is a potent modulator of nuclear receptor subfamily 2 group F member 6 (NR2F6). NR2F6 modulator-1 can be used to study immune regulation and modulation of tumour stem cell activity. purity: 97%