Learn More
Medchemexpress LLC NR2F6 modulator-1 | 904449-84-9 | 99.5% | 419.45 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY147228
Encompass_Preferred
NR2F6 modulator-1 is a potent nuclear receptor subfamily 2, group F, member 6 (NR2F6) modulator. This compound can be utilized for researching immune modulation and the modulation of cancer stem cell activity.
  • Potent NR2F6 modulator
  • Can be used for researching immune modulation
  • Useful for modulation of cancer stem cell activity
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO (125 mg/mL)
  • In vivo solubility in 10% DMSO, 90% corn oil (≥ 2.08 mg/mL)
Catalog No. 50-003-63262