Learn More
Aviva Systems Biology Corp NR3C1 Antibody - N-terminal region (ARP45593_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45593_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR3C1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NR3C1. Alternate names: GCCR, GCR, GR, GRLApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-59056