Learn More
Aviva Systems Biology Corp NR3C2 Antibody - middle region (ARP45599_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45599_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NR3C2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human NR3C2. Alternate names: MCR, MGC133092, MLR, MR, NR3C2VITApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59060