Learn More
Aviva Systems Biology Corp NR3C2 Antibody - N-terminal region (ARP31008_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31008_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR3C2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NR3C2. Alternate names: MCR, MGC133092, MLR, MR, NR3C2VITApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54175