Learn More
Aviva Systems Biology Corp NR4A1 Antibody - N-terminal region (ARP31941_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31941_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NR4A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NR4A1. Alternate names: HMR, N10, TR3, NP10, GFRP1, NAK-1, NGFIB, NUR77ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54384