Learn More
Aviva Systems Biology Corp NR4A2 Antibody - C-terminal region (ARP32730_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32730_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR4A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NR4A2. Alternate names: HZF-3, NOT, NURR1, RNR1, TINURApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54730