Learn More
Aviva Systems Biology Corp NR5A1 Antibody - C-terminal region (ARP37471_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37471_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NR5A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: ELP, SF1, SF-1, Ad4BP, ELP-3, Ftzf1, Ftz-F1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-56373