Learn More
Aviva Systems Biology Corp NR5A1 Antibody - middle region (ARP45620_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45620_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR5A1. It was validated on Western Blot. . Alternate names: AD4BP, ELP, FTZ1, FTZF1, POF7, SF-1, SF1, SPGF8, SRXY3ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-285-209