Learn More
Aviva Systems Biology Corp NR5A1 Antibody - middle region (ARP31834_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31834_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR5A1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human NR5A1. Alternate names: AD4BP, ELP, FTZ1, FTZF1, SF-1, SF1, POF7, SPGF8, SRXY3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54334