Learn More
Cayman Chemical NR5A1C247SC412S mutnth 100ug
SDP
Catalog No. 502945120
Encompass_Preferred
Source: Recombinant human N-terminal His-tagged HR5A1 (C247S, C412S mutant) expressed in E. coli • Amino acids: 218-461 • MW: 31.01 kDa
Catalog No. 50-294-5120 Supplier Cayman Chemical Supplier No. 41072100 UG
Source: Recombinant human N-terminal His-tagged HR5A1 (C247S, C412S mutant) expressed in E. coli • Amino acids: 218-461 • MW: 31.01 kDa