Learn More
Aviva Systems Biology Corp NR5A2 Antibody - N-terminal region (ARP37408_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37408_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nr5a2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human Nr5a2. Alternate names: AU020803, D1Ertd308e, Ftf, LRH-1ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-626