Learn More
Aviva Systems Biology Corp NR6A1 Antibody - N-terminal region (ARP32554_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32554_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR6A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NR6A1. Alternate names: GCNF, GCNF1, NR61, RTRApplicationWB. Species reactivity: Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54653