Learn More
Aviva Systems Biology Corp NR6A1 Antibody - middle region (ARP45640_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45640_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR6A1. It was validated on Western Blot. NR6A1,nuclear receptor subfamily 6, group A, member 1,RTR; GCNF; NR61; GCNF1; NR6A1,nuclear receptor subfamily 6 group A member 1,nuclear receptor subfamily 6 group A member 1,hRTR,hGCNF,OTTHUMP00000215858,germ cell nuclear factor,retinoid receptor-related testis-specific receptor,retinoic acid receptor-related testis-associated receptorThe immunogen is a synthetic peptide directed towards the middle region of human NR6A1. Alternate names: GCNF, GCNF1, NR61, RTRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-285-212