Learn More
Medchemexpress LLC NRAS Human N-His 5ug
SDP
Catalog No. 5000179215
Encompass_Preferred
NRAS protein is a member of the Ras family that binds GDP/GTP and has intrinsic GTPase activity This property enables NRAS to actively regulate cellular processes by cycling between a GDP-bound inactive state and a GTP-bound active state NRAS Protein Human (N-His) is the recombinant human-derived NRAS protein expressed by E coli with N-His labeled tag
Catalog No. 50-001-79215 Supplier Medchemexpress LLC Supplier No. HYP73741A5UG
NRAS protein is a member of the Ras family that binds GDP/GTP and has intrinsic GTPase activity This property enables NRAS to actively regulate cellular processes by cycling between a GDP-bound inactive state and a GTP-bound active state NRAS Protein Human (N-His) is the recombinant human-derived NRAS protein expressed by E coli with N-His labeled tag