Learn More
Sino Biological Recombinant Human KRAS Protein (Mature form, His Tag), Active
Catalog No. 502573298
Encompass_Preferred
Recombinant human KRAS (2-186) was expressed in E.coli cells using an N-terminal His tag.
Catalog No. 50-257-3298 Supplier Sino Biological Supplier No. R106310H500
Recombinant human KRAS (2-186) was expressed in E.coli cells using an N-terminal His tag.