Learn More
Aviva Systems Biology Corp NRBF2 Antibody - N-terminal region (ARP42660_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42660_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NRBF2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human NRBF2. Alternate names: COPR1, COPR2, DKFZp564C1664, FLJ30395, NRBF-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-017