Learn More
Aviva Systems Biology Corp NRBP1 Antibody - N-terminal region (ARP54949_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54949_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NRBP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human NRBP1. Alternate names: BCON3, FLJ27109, FLJ35541, MADM, MUDPNP, NRBPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-981