Learn More
Aviva Systems Biology Corp NRBP2 Antibody - middle region (ARP58823_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58823_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NRBP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NRBP2. Alternate names: DKFZp434P086, MGC138699, TRG16, pp9320ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-62437