Learn More
Aviva Systems Biology Corp NRCAM Antibody - N-terminal region (ARP44832_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44832_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NRCAM. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NRCAM. Alternate names: KIAA0343, MGC138845, MGC138846ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58910